Assay Details
Gene Symbol: Nrm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212508.2 | NP_997673.1 | 2 - 3 | 385 | 76 | |
XM_008772732.1 | XP_008770954.1 | 2 - 3 | 1317 | 76 | ||
GenBank mRNA | BC098637.1 | - | 2 - 3 | 346 | 76 | |
BP504258.1 | - | 2 - 3 | 391 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
