Assay Details
Gene Symbol: Abcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109883.2 | NP_001103353.1 | 21 - 22 | 2252 | 64 | |
GenBank mRNA | AF293383.1 | - | 20 - 21 | 2148 | 64 | |
CV114142.1 | - | 6 - 7 | 682 | 64 | ||
EV771702.1 | - | 4 - 5 | 305 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
