Assay Details
Gene Symbol: Atf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100895.1 | NP_001094365.1 | 1 - 2 | 261 | 158 | |
GenBank mRNA | BC091222.1 | - | 1 - 2 | 235 | 158 | |
BC105853.1 | - | 1 - 2 | 258 | 158 | ||
BC168224.1 | - | 1 - 2 | 262 | 158 | ||
CO565801.1 | - | 261 | 158 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001100895.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
