Assay Details
Gene Symbol: Atf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100895.1 | NP_001094365.1 | 2 - 3 | 356 | 119 | |
GenBank mRNA | BC091222.1 | - | 2 - 3 | 330 | 119 | |
BC105853.1 | - | 2 - 3 | 353 | 119 | ||
BC168224.1 | - | 2 - 3 | 357 | 119 | ||
CO565801.1 | - | 356 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
