Assay Details
Gene Symbol: Atf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100895.1 | NP_001094365.1 | 6 - 7 | 932 | 128 | |
GenBank mRNA | BC091222.1 | - | 6 - 7 | 906 | 128 | |
BC105853.1 | - | 6 - 7 | 929 | 128 | ||
BC168224.1 | - | 7 - 8 | 984 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
transcription by RNA polymerase II
positive regulation of neuron projection development
response to organic cyclic compound
response to cobalt ion
cellular protein-containing complex assembly
positive regulation of DNA replication
positive regulation of transcription by RNA polymerase II
RNA polymerase II transcription regulatory region sequence-specific DNA binding
DNA-binding transcription factor activity, RNA polymerase II-specific
DNA-binding transcription activator activity, RNA polymerase II-specific
DNA binding
sequence-specific DNA binding
protein-containing complex binding
protein heterodimerization activity
transcription by RNA polymerase II
positive regulation of neuron projection development
response to organic cyclic compound
response to cobalt ion
cellular protein-containing complex assembly
positive regulation of DNA replication
positive regulation of transcription by RNA polymerase II
RNA polymerase II transcription regulatory region sequence-specific DNA binding
DNA-binding transcription factor activity, RNA polymerase II-specific
DNA-binding transcription activator activity, RNA polymerase II-specific
DNA binding
sequence-specific DNA binding
protein-containing complex binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P166513 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P633046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35787 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P166513 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P633046 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P633046 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P166513 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P166513 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P633046 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P166513 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P718305 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83956 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10899254 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899253 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899252 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899251 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899249 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899256 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899250 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899255 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10899248 | - | - |
Rat Genome 230 2.0 Array | 1389623_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389623_at | - | - |
Affymetrix HT Rat Focus Array | 1389623_at | - | - |
Affymetrix HT Rat Focus Array | 1389623_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700013561.rn.1 | TC0700001763.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700013559.rn.1 | TC0700001763.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700013560.rn.1 | TC0700001763.rn.1 | RTA TC |
Rat Expression Set 230 | 1389623_PM_at | - | - |
Rat Expression Set 230 | 1389623_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10899248 | - | - |
Rat Genome U34 Set | rc_AA849092_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21919370 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356841 | - | - |
