Assay Details
Gene Symbol: Bin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012223.1 | NP_001012223.1 | 3 - 4 | 262 | 84 | |
XM_008765773.1 | XP_008763995.1 | 3 - 4 | 289 | 84 | ||
GenBank mRNA | BC079406.1 | - | 3 - 4 | 262 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008765773.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
