Assay Details
Gene Symbol: Fkbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013105.2 | NP_001013123.2 | 2 - 3 | 288 | 106 | |
GenBank mRNA | BC084709.1 | - | 2 - 3 | 266 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
