Assay Details
Gene Symbol: Fkbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013105.2 | NP_001013123.2 | 4 - 5 | 403 | 93 | |
XM_006257344.2 | XP_006257406.1 | 3 - 4 | 1308 | 93 | ||
XM_008765695.1 | XP_008763917.1 | 3 - 4 | 306 | 93 | ||
XM_008765696.1 | XP_008763918.1 | 3 - 4 | 318 | 93 | ||
XM_008765697.1 | XP_008763919.1 | 3 - 4 | 295 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008765695.1, XM_008765696.1, XM_008765697.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
