Assay Details
Gene Symbol: Cers5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108993.1 | NP_001102463.1 | 2 - 3 | 443 | 98 | |
XM_006257374.2 | XP_006257436.1 | 2 - 3 | 426 | 98 | ||
GenBank mRNA | BC166794.1 | - | 2 - 3 | 325 | 7937721 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
