Assay Details
Gene Symbol: Cers5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108993.1 | NP_001102463.1 | 6 - 7 | 778 | 79 | |
XM_006257374.2 | XP_006257436.1 | 6 - 7 | 761 | 79 | ||
XM_006257375.2 | XP_006257437.1 | 4 - 5 | 620 | 79 | ||
GenBank mRNA | BC166794.1 | - | 6 - 7 | 660 | 7937721 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
