Assay Details
Gene Symbol: Sp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012655.2 | NP_036787.2 | 2 - 3 | 200 | 96 | |
GenBank mRNA | D12768.1 | - | 2 - 3 | 210 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
