Assay Details
Gene Symbol: Pcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013223.1 | NP_001013241.1 | 4 - 5 | 356 | 124 | |
XM_006242417.2 | XP_006242479.1 | 4 - 5 | 366 | 124 | ||
XM_006242418.2 | XP_006242480.1 | 4 - 5 | 366 | 124 | ||
XM_006242419.2 | XP_006242481.1 | 4 - 5 | 288 | 124 | ||
XM_006242420.2 | XP_006242482.1 | 4 - 5 | 366 | 124 | ||
XM_006242421.2 | XP_006242483.1 | 4 - 5 | 366 | 124 | ||
XM_006242422.2 | XP_006242484.1 | 4 - 5 | 366 | 124 | ||
XM_006242423.2 | XP_006242485.1 | 4 - 5 | 366 | 124 | ||
XM_006242424.2 | XP_006242486.1 | 4 - 5 | 786 | 124 | ||
XM_006242425.2 | XP_006242487.1 | 4 - 5 | 366 | 124 | ||
XM_006242426.2 | XP_006242488.1 | 4 - 5 | 366 | 124 | ||
XM_006242427.2 | XP_006242489.1 | 4 - 5 | 366 | 124 | ||
XM_008765770.1 | XP_008763992.1 | 4 - 5 | 288 | 124 | ||
XM_008765771.1 | XP_008763993.1 | 4 - 5 | 288 | 124 | ||
GenBank mRNA | BC078906.1 | - | 4 - 5 | 356 | 7112287 | 124 |
BC078909.1 | - | 3 - 4 | 195 | 7127139 | 124 | |
BC097410.1 | - | 5 - 6 | 471 | 7454176 | 124 | |
FQ216778.1 | - | 4 - 5 | 257 | 124 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
