Assay Details
Gene Symbol: Pcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013223.1 | NP_001013241.1 | 6 - 7 | 598 | 99 | |
XM_006242417.2 | XP_006242479.1 | 6 - 7 | 608 | 99 | ||
XM_006242418.2 | XP_006242480.1 | 6 - 7 | 608 | 99 | ||
XM_006242419.2 | XP_006242481.1 | 6 - 7 | 530 | 99 | ||
XM_006242420.2 | XP_006242482.1 | 6 - 7 | 608 | 99 | ||
XM_006242421.2 | XP_006242483.1 | 6 - 7 | 608 | 99 | ||
XM_006242422.2 | XP_006242484.1 | 6 - 7 | 608 | 99 | ||
XM_006242423.2 | XP_006242485.1 | 6 - 7 | 608 | 99 | ||
XM_006242424.2 | XP_006242486.1 | 6 - 7 | 1028 | 99 | ||
XM_006242425.2 | XP_006242487.1 | 6 - 7 | 608 | 99 | ||
XM_006242426.2 | XP_006242488.1 | 6 - 7 | 608 | 99 | ||
XM_006242427.2 | XP_006242489.1 | 6 - 7 | 608 | 99 | ||
XM_008765770.1 | XP_008763992.1 | 6 - 7 | 530 | 99 | ||
XM_008765771.1 | XP_008763993.1 | 6 - 7 | 530 | 99 | ||
GenBank mRNA | BC078906.1 | - | 6 - 7 | 598 | 99 | |
BC078909.1 | - | 5 - 6 | 437 | 99 | ||
BC097410.1 | - | 7 - 8 | 713 | 99 | ||
FQ216778.1 | - | 6 - 7 | 499 | 99 | ||
FQ231388.1 | - | 2 - 3 | 151 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
