Assay Details
Gene Symbol: Tmbim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019381.2 | NP_062254.2 | 4 - 5 | 371 | 150 | |
XM_006257323.2 | XP_006257385.1 | 4 - 5 | 513 | 150 | ||
XM_008765670.1 | XP_008763892.1 | 5 - 6 | 471 | 150 | ||
GenBank mRNA | AY321320.1 | - | 6 - 7 | 509 | 150 | |
AY325130.1 | - | 6 - 7 | 509 | 150 | ||
AY325243.1 | - | 6 - 7 | 509 | 150 | ||
BC058478.1 | - | 4 - 5 | 377 | 6888036 | 150 | |
FQ217052.1 | - | 4 - 5 | 370 | 150 | ||
FQ218664.1 | - | 4 - 5 | 373 | 150 | ||
FQ226858.1 | - | 4 - 5 | 400 | 150 | ||
FQ229652.1 | - | 4 - 5 | 372 | 150 | ||
FQ230608.1 | - | 4 - 5 | 400 | 150 | ||
FQ231817.1 | - | 4 - 5 | 378 | 150 | ||
X75855.1 | - | 4 - 5 | 400 | 147 | ||
X75856.1 | - | 3 - 4 | 284 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
