Assay Details
Gene Symbol: Rarg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135249.1 | NP_001128721.1 | 3 - 4 | 671 | 134 | |
XM_008765785.1 | XP_008764007.1 | 3 - 4 | 635 | 134 | ||
XM_008765786.1 | XP_008764008.1 | 2 - 3 | 1163 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
