Assay Details
Gene Symbol: Cela1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012552.3 | NP_036684.1 | 3 - 4 | 227 | 101 | |
GenBank mRNA | FQ227752.1 | - | 3 - 4 | 249 | 101 | |
V01234.1 | - | 3 - 4 | 248 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
