Assay Details
Gene Symbol: Cela1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012552.3 | NP_036684.1 | 4 - 5 | 350 | 136 | |
GenBank mRNA | FQ227752.1 | - | 4 - 5 | 372 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
