Assay Details
Gene Symbol: Cela1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012552.3 | NP_036684.1 | 6 - 7 | 631 | 93 | |
GenBank mRNA | FQ227752.1 | - | 6 - 7 | 653 | 93 | |
V01234.1 | - | 6 - 7 | 652 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
