Assay Details
Gene Symbol: Vdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017058.1 | NP_058754.1 | 7 - 8 | 837 | 93 | |
GenBank mRNA | J04147.1 | - | 7 - 8 | 837 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
