Assay Details
Gene Symbol: Prkab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022627.2 | NP_072149.1 | 4 - 5 | 540 | 177 | |
XM_006233011.2 | XP_006233073.1 | 5 - 6 | 741 | 177 | ||
XM_006233012.2 | XP_006233074.1 | 5 - 6 | 641 | 177 | ||
GenBank mRNA | AF182717.1 | - | 4 - 5 | 542 | 177 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
