Assay Details
Gene Symbol: Otud7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107697.1 | NP_001101167.1 | 2 - 3 | 448 | 127 | |
XM_006232957.2 | XP_006233019.1 | 2 - 3 | 217 | 127 | ||
XM_006232958.2 | XP_006233020.1 | 1 - 2 | 159 | 127 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006232958.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
