Assay Details
Gene Symbol: Otud7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107697.1 | NP_001101167.1 | 4 - 5 | 855 | 117 | |
XM_006232957.2 | XP_006233019.1 | 4 - 5 | 624 | 117 | ||
XM_006232958.2 | XP_006233020.1 | 3 - 4 | 432 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
