Assay Details
Gene Symbol: Psmd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031331.1 | NP_112621.1 | 4 - 5 | 440 | 98 | |
XM_006232924.2 | XP_006232986.1 | 4 - 5 | 445 | 98 | ||
GenBank mRNA | AB017188.1 | - | 4 - 5 | 440 | 98 | |
AF175575.1 | - | 4 - 5 | 381 | 98 | ||
BC060559.1 | - | 4 - 5 | 390 | 6891370 | 98 | |
FQ209672.1 | - | 3 - 4 | 242 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
