Assay Details
Gene Symbol: Nudt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109340.1 | NP_001102810.1 | 1 - 2 | 221 | 64 | |
XM_008761327.1 | XP_008759549.1 | 1 - 2 | 1995 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
