Assay Details
Gene Symbol: Vangl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109584.1 | NP_001103054.1 | 3 - 4 | 418 | 60 | |
XM_006233054.2 | XP_006233116.1 | 3 - 4 | 459 | 60 | ||
XM_006233055.2 | XP_006233117.1 | 3 - 4 | 458 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
