Assay Details
Gene Symbol: Vangl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109584.1 | NP_001103054.1 | 7 - 8 | 1536 | 57 | |
XM_006233054.2 | XP_006233116.1 | 8 - 9 | 1616 | 57 | ||
XM_006233055.2 | XP_006233117.1 | 8 - 9 | 1579 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P055472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107925 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P055472 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P055472 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P055472 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57780 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20998 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10825515 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825518 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825517 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825514 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825519 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825520 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825516 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825513 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10825512 | - | - |
Rat Genome 230 2.0 Array | 1394435_at | - | - |
Rat Genome 230 2.0 Array | 1394435_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1394435_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1394435_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200029904.rn.1 | TC0200004343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200029905.rn.1 | TC0200004343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200029908.rn.1 | TC0200004343.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200029906.rn.1 | TC0200004343.rn.1 | RTA TC |
Rat Expression Set 230 | 1394435_at | - | - |
Rat Expression Set 230 | 1394435_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825512 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21672779 | - | - |
