Assay Details
Gene Symbol: Gabpb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232936.2 | XP_006232998.1 | 5 - 6 | 1330 | 117 | |
XM_227439.7 | XP_227439.6 | 5 - 6 | 923 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
