Assay Details
Gene Symbol: Bnipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128187.2 | NP_001121659.1 | 1 - 2 | 88 | 129 | |
GenBank mRNA | BC166747.1 | - | 1 - 2 | 88 | 129 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC166747.1, NM_001128187.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
