Assay Details
Gene Symbol: Bnipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128187.2 | NP_001121659.1 | 2 - 3 | 178 | 88 | |
XM_006232889.2 | XP_006232951.1 | 1 - 2 | 517 | 88 | ||
GenBank mRNA | BC166747.1 | - | 2 - 3 | 178 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
