Assay Details
Gene Symbol: Bnipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128187.2 | NP_001121659.1 | 9 - 10 | 1069 | 93 | |
XM_006232889.2 | XP_006232951.1 | 8 - 9 | 1408 | 93 | ||
GenBank mRNA | BC166747.1 | - | 9 - 10 | 1069 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
