Assay Details
Gene Symbol: Ampd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138876.1 | NP_620231.1 | 6 - 7 | 774 | 85 | |
XM_006233060.2 | XP_006233122.1 | 5 - 6 | 851 | 85 | ||
XM_008761369.1 | XP_008759591.1 | 5 - 6 | 774 | 85 | ||
GenBank mRNA | J02811.1 | - | 6 - 7 | 774 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
