Assay Details
Gene Symbol: Txnip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008767.1 | NP_001008767.1 | 6 - 7 | 1177 | 152 | |
GenBank mRNA | BC088411.1 | - | 6 - 7 | 1177 | 152 | |
U30789.1 | - | 6 - 7 | 1962 | 152 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
