Assay Details
Gene Symbol: Oaz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101018.1 | NP_001094488.1 | 3 - 4 | 526 | 78 | |
GenBank mRNA | AW529628.1 | - | 2 - 3 | 78 | ||
DQ431008.1 | - | 3 - 4 | 526 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
