Assay Details
Gene Symbol: Polr3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012081.1 | NP_001012081.1 | 8 - 9 | 1312 | 100 | |
XM_006232967.1 | XP_006233029.1 | 8 - 9 | 1519 | 100 | ||
XM_008761310.1 | XP_008759532.1 | 8 - 9 | 1242 | 100 | ||
XM_008761311.1 | XP_008759533.1 | 8 - 9 | 1519 | 100 | ||
XM_008761312.1 | XP_008759534.1 | 7 - 8 | 1333 | 100 | ||
GenBank mRNA | BC083667.1 | - | 8 - 9 | 1312 | 7135563 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
MicroArray Probes:
