Assay Details
Gene Symbol: Rbm8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271138.1 | NP_001258067.1 | 2 - 3 | 173 | 126 | |
GenBank mRNA | DQ359102.1 | - | 2 - 3 | 125 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
