Assay Details
Gene Symbol: Rbm8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271138.1 | NP_001258067.1 | 5 - 6 | 531 | 61 | |
XM_006232956.2 | XP_006233018.1 | 5 - 6 | 529 | 61 | ||
GenBank mRNA | DQ359102.1 | - | 5 - 6 | 483 | 61 | |
FQ210627.1 | - | 5 - 6 | 504 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
