Assay Details
Gene Symbol: Fam46c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014041.1 | NP_001014063.1 | 1 - 2 | 237 | 70 | |
GenBank mRNA | BC083570.1 | - | 1 - 2 | 237 | 7111947 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC083570.1, NM_001014041.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
