Assay Details
Gene Symbol: Vps72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107695.1 | NP_001101165.1 | 2 - 3 | 335 | 59 | |
XM_008761302.1 | XP_008759524.1 | 2 - 3 | 367 | 59 | ||
GenBank mRNA | BC161999.1 | - | 2 - 3 | 323 | 9036078 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
