Assay Details
Gene Symbol: Igsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106455.1 | NP_001099925.1 | 3 - 4 | 842 | 71 | |
XM_006233043.2 | XP_006233105.1 | 4 - 5 | 1585 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
