Assay Details
Gene Symbol: Gdap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013201.1 | NP_001013219.1 | 12 - 13 | 1517 | 155 | |
XM_006233032.2 | XP_006233094.1 | 13 - 14 | 1615 | 155 | ||
GenBank mRNA | BC082000.1 | - | 12 - 13 | 1517 | 155 | |
FQ211326.1 | - | 14 - 15 | 1698 | 155 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P354053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138555 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P354053 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P354053 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P354053 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P354053 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8018 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48439 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4362 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23310 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10817838 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10817835 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10817841 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10817836 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10817843 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10817839 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10817833 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10817840 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10817842 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10817844 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10817832 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10817834 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10817831 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10817837 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10817830 | - | - |
Rat Genome 230 2.0 Array | 1379377_PM_at | - | - |
Rat Genome 230 2.0 Array | 1379377_at | - | - |
Affymetrix HT Rat Focus Array | 1379377_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379377_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200012520.rn.1 | TC0200001830.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200012519.rn.1 | TC0200001830.rn.1 | RTA TC |
Rat Expression Set 230 | 1379377_at | - | - |
Rat Expression Set 230 | 1379377_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10817830 | - | - |
Rat Genome U34 Set | rc_AI178884_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22061774 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351978 | - | - |
