Assay Details
Gene Symbol: Cab39l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011917.1 | NP_001011917.1 | 1 - 2 | 145 | 63 | |
XM_006252098.2 | XP_006252160.1 | 2 - 3 | 249 | 63 | ||
XM_006252099.2 | XP_006252161.1 | 2 - 3 | 249 | 63 | ||
XM_006252100.2 | XP_006252162.1 | 2 - 3 | 249 | 63 | ||
XM_006252101.2 | XP_006252163.1 | 2 - 3 | 249 | 63 | ||
XM_008770742.1 | XP_008768964.1 | 3 - 4 | 342 | 63 | ||
GenBank mRNA | BC083684.1 | - | 1 - 2 | 145 | 7188949 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC083684.1, NM_001011917.1, XM_006252098.2, XM_006252099.2, XM_006252100.2, XM_006252101.2, XM_008770742.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
