Assay Details
Gene Symbol: Chrna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133420.1 | NP_596911.1 | 5 - 6 | 689 | 60 | |
XM_008770735.1 | XP_008768957.1 | 5 - 6 | 531 | 60 | ||
GenBank mRNA | AY574253.1 | - | 4 - 5 | 385 | 60 | |
L10077.1 | - | 5 - 6 | 689 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
