Assay Details
Gene Symbol: Clu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053021.2 | NP_444180.2 | 3 - 4 | 391 | 110 | |
XM_006252094.2 | XP_006252156.1 | 2 - 3 | 269 | 110 | ||
GenBank mRNA | BC061534.1 | - | 2 - 3 | 170 | 110 | |
FQ209467.1 | - | 2 - 3 | 167 | 110 | ||
FQ209498.1 | - | 2 - 3 | 168 | 110 | ||
FQ209623.1 | - | 2 - 3 | 166 | 110 | ||
FQ211071.1 | - | 2 - 3 | 166 | 110 | ||
FQ213433.1 | - | 2 - 3 | 165 | 110 | ||
FQ214098.1 | - | 2 - 3 | 211 | 110 | ||
FQ218237.1 | - | 2 - 3 | 166 | 110 | ||
FQ218812.1 | - | 2 - 3 | 166 | 110 | ||
FQ219030.1 | - | 2 - 3 | 166 | 110 | ||
M16975.1 | - | 3 - 4 | 391 | 110 | ||
M64723.1 | - | 2 - 3 | 156 | 110 | ||
X13231.1 | - | 2 - 3 | 155 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
