Assay Details
Gene Symbol: Clu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053021.2 | NP_444180.2 | 5 - 6 | 707 | 98 | |
XM_006252094.2 | XP_006252156.1 | 4 - 5 | 585 | 98 | ||
GenBank mRNA | BC061534.1 | - | 4 - 5 | 486 | 98 | |
FQ209467.1 | - | 4 - 5 | 483 | 98 | ||
FQ209498.1 | - | 4 - 5 | 484 | 98 | ||
FQ209623.1 | - | 4 - 5 | 482 | 98 | ||
FQ211071.1 | - | 4 - 5 | 481 | 98 | ||
FQ213433.1 | - | 4 - 5 | 481 | 98 | ||
FQ214098.1 | - | 4 - 5 | 527 | 98 | ||
FQ218237.1 | - | 4 - 5 | 482 | 98 | ||
FQ218812.1 | - | 4 - 5 | 482 | 98 | ||
FQ219030.1 | - | 4 - 5 | 482 | 98 | ||
M16975.1 | - | 5 - 6 | 707 | 98 | ||
M64723.1 | - | 4 - 5 | 472 | 98 | ||
X13231.1 | - | 4 - 5 | 471 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
