Assay Details
Gene Symbol: Clu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053021.2 | NP_444180.2 | 7 - 8 | 1225 | 60 | |
XM_006252094.2 | XP_006252156.1 | 6 - 7 | 1103 | 60 | ||
GenBank mRNA | AF314657.1 | - | 3 - 4 | 241 | 60 | |
BC061534.1 | - | 6 - 7 | 1004 | 60 | ||
FQ209467.1 | - | 6 - 7 | 1004 | 60 | ||
FQ211071.1 | - | 6 - 7 | 998 | 60 | ||
FQ213433.1 | - | 6 - 7 | 1007 | 60 | ||
FQ214098.1 | - | 6 - 7 | 1045 | 60 | ||
FQ218812.1 | - | 6 - 7 | 1002 | 60 | ||
FQ219030.1 | - | 6 - 7 | 999 | 60 | ||
M16975.1 | - | 7 - 8 | 1225 | 60 | ||
M64723.1 | - | 6 - 7 | 990 | 60 | ||
X13231.1 | - | 6 - 7 | 989 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
