Assay Details
Gene Symbol: Tnfrsf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044229.2 | NP_001037694.1 | 6 - 7 | 741 | 64 | |
GenBank mRNA | CO396223.1 | - | 2 - 3 | 287 | 64 | |
DQ466087.1 | - | 6 - 7 | 741 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
