Assay Details
Gene Symbol: Sacs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252167.2 | XP_006252229.1 | 4 - 5 | 1004 | 146 | |
XM_006252168.2 | XP_006252230.1 | 3 - 4 | 532 | 146 | ||
XM_006252169.2 | XP_006252231.1 | 4 - 5 | 1004 | 146 | ||
XM_008770792.1 | XP_008769014.1 | 4 - 5 | 1004 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
