Assay Details
Gene Symbol: Pinx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006252181.2 | XP_006252243.1 | 5 - 6 | 1298 | 148 | |
GenBank mRNA | BC158553.1 | - | 5 - 6 | 494 | 148 | |
FQ232228.1 | - | 4 - 5 | 283 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
