Assay Details
Gene Symbol: Sgcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006993.1 | NP_001006994.1 | 3 - 4 | 385 | 159 | |
GenBank mRNA | BC083748.1 | - | 3 - 4 | 385 | 7192349 | 159 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
