Assay Details
Gene Symbol: Aadat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017193.1 | NP_058889.1 | 5 - 6 | 772 | 99 | |
XM_006253067.2 | XP_006253129.1 | 5 - 6 | 860 | 99 | ||
GenBank mRNA | BC078864.1 | - | 5 - 6 | 768 | 99 | |
Z50144.1 | - | 5 - 6 | 772 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
